Chloromaleic acid anhydride
Fornecedor: Apollo Scientific
|
Perigo
|
OR30483-1G
OR30483-250MG
APOSOR30483-1GEA
0
EUR
APOSOR30483-1G
APOSOR30483-250MG
Chloromaleic acid anhydride
Chloromaleic acid anhydride
Fórmula:
C₄HClO₃ Temperatura de Armazenamento: Ambiente |
Número MDL :
MFCD00210253 Número CAS: 96-02-6 UN: 2923 ADR: 8+6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...