Print…
2-Fluoro-4-nitrophenyl 2-azido-2-deoxy-β-D-galactopyranoside 95%
Fornecedor: Apollo Scientific
BICL4311-10MG BICL4311-25MG BICL4311-50MG
APOSBICL4311-10MGEA 364 EUR
APOSBICL4311-10MG APOSBICL4311-25MG APOSBICL4311-50MG
2-Fluoro-4-nitrophenyl 2-azido-2-deoxy-β-D-galactopyranoside 95%
2-Fluoro-4-nitrophenyl 2-azido-2-deoxy-β-D-galactopyranoside