Print…
VWR® Burettes, with PTFE Needle Valve Stopcock
Fornecedor: VWR Collection
612-6688EA 81.1 EUR
612-6688 612-6689 612-6690
VWR® Burettes, with PTFE Needle Valve Stopcock
Buretas
Borosilicate 3.3 glass, class AS.
  • High chemical and thermal resistance
  • Schellbach stripe for accurate meniscus reading
  • Stopcock with PTFE needle valve
  • High contrast blue enamel graduations
  • Batch certificate included
To avoid material damaging friction between the burette and the stopcock and to ensure tightness joint, grease is necessary when working with burettes with glass stopcock. After use the joint grease must be removed as the burette tip could become contaminated. This step is not necessary when working with burettes with a PTFE needle valve stopcock.
Certificações: DIN EN ISO 385