Restriction enzymes, EcoRI, HC
Fornecedor: Promega
R4014
R4017
PROMR4014EA
178
EUR
PROMR4014
PROMR401750KU
Restriction enzymes, EcoRI, HC
Reagentes de ácidos nucleicos
Restriction Enzymes
These EcoRI restriction enzyme recognises and cut G˅AATT C and C TTAA^G at 37 °C in its own unique buffer.
- Capable of digesting DNA in 15 minutes or less
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
- Source: Escherichia coli RY 13
Storage buffer: 10 mM Tris-HCl (pH 7.4), 400 mM NaCl, 0,1 mM EDTA, 1 mM DTT, 0,15% Triton® X-100, 0,5 mg/ml BSA, 50% glycerol.
Fornecimento: Kit includes 1×1 ml buffer H 10X buffer, 1×750 µl bovine serum albumin (acetylated) and 1×5,000 units EcoRI.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...