Print…
DURAN® Autoclave Bottle, for Use with 45 mm Push-On Rubber Cap
Fornecedor: DWK Life Sciences
219946305 292063002
215-4638EA 40.3 EUR
215-4638 217-0899
DURAN® Autoclave Bottle, for Use with 45 mm Push-On Rubber Cap
Frascos Frascos de Vidro
Moulded borosilicate 3.3 glass bottle for autoclaving and storing sterile liquids, compatible with 45 mm self-venting rubber cap.
  • Fully compatible with 45 mm self-venting, rubber sealing cap NC 003
  • Autoclavable at 121 or 134 °C
  • Borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack
  • High-purity, clear, USP/EP/JP Type I glass
  • Retrace code for manufacturing lot tracability
  • Fired-on, highly durable white print
  • Easy-to-read double graduated scales (500 - 1800 ml, 100 ml intervals) to show either liquid volume contained or volume poured out
Applications: stea /autoclave sterilisation of water and media, storage of sterile liquids and media.
Certificações: DIN ISO 3585.
Glass Type I according to the requirements of the current United States pharmacopoeia USP <660> Containers – Glass.
Glass Type I according to the requirements of the current European pharmacopoeia EP (3.2.1) Glass Containers for Pharmaceutical Use.
Glass Type I according to the requirements of the current Japanese pharmacopoeia JP <7.01> Test for Glass Containers for injections.
The calibration results of volumetric instruments with this certificate refer to one calibration batch.
This batch certificate confirms that ASTM Class A error limits required by USP are met. The certificate records the mean value, the standard deviation of the batch and the date of issue.