Print…
Multi-position hotplates, MultiHeat D
Fornecedor: SUMINISTROS GRUPO ESPER S L
10000-01043 10000-01044 10000-01047
453-0077EA 2520 EUR
453-0077 453-0078 453-0080
Multi-position hotplates, MultiHeat D
Aquecimentos Placas de aquecimento
Polished aluminium, digital multi-position hotplates, with microprocessor control. Modules consist of heating units in a single platform, with a single electrical connection. Each position works independently.
  • Digital display LCD backlighted indicating the selected and actual temperature
  • Membrane push-button control
Models with 5 or 9 square plates: user friendly interface to adjust the temperature at each position and the selected parameters are memorised, even if the instrument is switched off.
Models with 3 round plates: adjustable temperature, controlled by means of an integrated PT100 sensor. 'ARA V2' fuzzy logic algorithm for precise control of the temperature, to be used with external sensor without exceeding reference temperature (even at 30 °C). Timer 0 to 120 min. with audible alarm. Indicator on display for high plate temperature. Automatic restart after unexpected shut down keeps the same parameters as before an incident and removable plates.