Print…
Baths for oil testing, BCA6E and BCA9E
Fornecedor: SUMINISTROS GRUPO ESPER S L
10000-01133 10000-01135
453-0084EA 1870 EUR
453-0084 453-0085
Baths for oil testing, BCA6E and BCA9E
Blocos de aquecimento Blocos de aquecimento digitais
Digital thermostatic baths for oil testing, controlled by microprocessor. Aluminium block with six or nine holes, specially designed for edible oil testing.
  • Digital backlit LCD display showing the selected and actual temperature, and timer (0 to 120 min.)
  • Display shows high temperature on the tank, giving protection from burns
  • Automatic restart after unexpected shut down keeping the same parameters as before the incident. An incident sign appears on the display
  • Memory of the last settings
  • Special stainless steel alloy heater is resistant to corrosion and high temperatures
  • The unit will cut out if there is insufficient liquid or if the temperature exceeds the maximum temperature
  • Push-button Membrane control
Stainless steel inner tank and metallic housing are protected with epoxy paint which has high resistance to aggressive surroundings.
Certificações: IP Protection class: IP 52