Print…
Nickel ICP standards, (CRMs), Fluka™
Fornecedor: Honeywell Chemicals
E1040-100ML E1168-100ML
HONCE1040-100MLEA 93.84 EUR
HONCE1040-100ML HONCE1168-100ML
Nickel ICP standards, (CRMs), Fluka™
Padrões Cationic Standards Nickel Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificações: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).