Print…
ZnTAC24® (contains 7% Toluene at maximum)
Fornecedor: TCI
Perigo
Sinônimos: Oxo[hexa(trifluoroacetato)]tetrazinc and its trifluoroacetic acid adduct
Z0035-1G Z0035-5G
TCIAZ0035-1GEA 18 EUR
TCIAZ0035-1G TCIAZ0035-5G
ZnTAC24® (contains 7% Toluene at maximum)
ZnTAC24®
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4777a9bf0f857f98b4bba274a3b2ed3a1f69c67c&vid=4777a9bf0f857f98b4bba274a3b2ed3a1f69c67c&prid=web&clid=SAPDAM
Encomendar
Appearance White to Light gray to Light yellow powder to crystal
Zinc 24 to 28 %