Print…

Tetrahydropalmatine ≥98.0% (por HPLC, nitrogênio total)

Fornecedor: TCI

T3650-1G T3650-5G
TCIAT3650-1GEA 41 EUR
TCIAT3650-1G TCIAT3650-5G
Tetrahydropalmatine ≥98.0% (por HPLC, nitrogênio total)
Tetrahydropalmatine
Fórmula: C₂₁H₂₅NO₄
Número MDL : MFCD00214191
Número CAS: 2934-97-6

Encomendar

Especificações dos Resultados do Teste

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 148.0 to 152.0 deg-C
Specific rotation [a]20/D -3.0 to +3.0 deg (C=0.5, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR