Print…

Sodium bis(trifluoromethylsulphonyl)imide ≥98.0% (por análise titulométrica)

Fornecedor: TCI
Perigo

Sinônimos: Bis(trifluoromethanesulphonyl)imide sodium salt, Sodium triflimide, Sodium trifluoromethanesulphonimide, Sodium Bis(trifluoromethanesulphonyl)imide

S0989-1G S0989-5G
TCIAS0989-1GEA 48 EUR
TCIAS0989-1G TCIAS0989-5G
Sodium bis(trifluoromethylsulphonyl)imide ≥98.0% (por análise titulométrica)
Sodium bis(trifluoromethylsulphonyl)imide
Fórmula: C₂F₆NNaO₄S₂
Peso Molecular: 303.13 g/mol
Ponto de Fusão: 257°C
Número MDL : MFCD14156021
Número CAS: 91742-21-1
UN: 1759
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d897cda9ed5a9f2421027d993cbee4f65f5e8b8a&vid=d897cda9ed5a9f2421027d993cbee4f65f5e8b8a&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR