Print…

n-Pentylboronic acid (contains varying amounts of Anhydride)

Fornecedor: TCI
Atenção

P2267-5G P2267-25G
TCIAP2267-5GEA 57 EUR
TCIAP2267-5G TCIAP2267-25G
n-Pentylboronic acid (contains varying amounts of Anhydride)
n-Pentylboronic acid
Fórmula: C₅H₁₃BO₂
Ponto de Fusão: 94°C
Número MDL : MFCD01074651
Número CAS: 4737-50-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=026a7e2f4855e01005787a775ff9da97127b3110&vid=026a7e2f4855e01005787a775ff9da97127b3110&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 119.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR