Print…

Methyl-N-phenylcarbamate ≥98.0% (por GC)

Fornecedor: TCI

P1010-25G P1010-500G
TCIAP1010-25GEA 39 EUR
TCIAP1010-25G TCIAP1010-500G
Methyl-N-phenylcarbamate ≥98.0% (por GC)
Methyl-N-phenylcarbamate
Fórmula: C₈H₉NO₂
Ponto de ebulição: 126°C
Número MDL : MFCD00027552
Número CAS: 2603-10-3
EINECS: 220-014-2

Encomendar

Especificações dos Resultados do Teste

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Melting point 46.0 to 49.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR