Print…

Methoxy(dimethyl)octadecylsilane ≥90.0% (por GC)

Fornecedor: TCI
Atenção

Sinônimos: Methoxydimethyl(octadecyl)silane

M3011-25ML M3011-5ML
TCIAM3011-25MLEA 111 EUR
TCIAM3011-25ML TCIAM3011-5ML
Methoxy(dimethyl)octadecylsilane ≥90.0% (por GC)
Methoxy(dimethyl)octadecylsilane
Fórmula: C₂₁H₄₆OSi
Peso Molecular: 342.68 g/mol
Número MDL : MFCD00042920
Número CAS: 71808-65-6
EINECS: 276-039-4

Encomendar

Especificações dos Resultados do Teste

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR