Print…

Methylammonium tetrafluoroborate ≥98.0% (por base de azoto total)

Fornecedor: TCI
Perigo

Sinônimos: Methylamine tetrafluoroborate

M2990-1G M2990-5G
TCIAM2990-1GEA 50 EUR
TCIAM2990-1G TCIAM2990-5G
Methylammonium tetrafluoroborate ≥98.0% (por base de azoto total)
Methylammonium tetrafluoroborate
Fórmula: CH₆BF₄N
Peso Molecular: 118.87 g/mol
Número CAS: 42539-74-2
UN: 1759
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5c21407d9ea1aaf2c79308cb43ff461c146e9fa&vid=f5c21407d9ea1aaf2c79308cb43ff461c146e9fa&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 197.0 to 201.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR