Print…

4-Methyl-N-tosylbenzenesulphonamide ≥98.0% (por HPLC)

Fornecedor: TCI

Sinônimos: Bis(p-tolylsulphonyl)amine, Ditosylamine, Bis(p-toluenesulphonyl)amine, Bis(4-methylbenzenesulphonyl)amine

M2975-1G M2975-5G
TCIAM2975-1GEA 27 EUR
TCIAM2975-1G TCIAM2975-5G
4-Methyl-N-tosylbenzenesulphonamide ≥98.0% (por HPLC)
4-Methyl-N-tosylbenzenesulphonamide
Fórmula: C₁₄H₁₅NO₄S₂
Peso Molecular: 325.40 g/mol
Número MDL : MFCD00014917
Número CAS: 3695-00-9
EINECS: 223-016-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acc9a5ce618f23a648a0e0f41d1cdee5422e9b83&vid=acc9a5ce618f23a648a0e0f41d1cdee5422e9b83&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 171.0 to 175.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR