Print…

Manganesebis(trifluoromethylsulphonyl)imide ≥97.0% (por análise titulométrica)

Fornecedor: TCI
Perigo

Sinônimos: Manganese(II) triflimide, Manganese(II) Bis(trifluoromethanesulphonyl)imide, Manganese bis(trifluoromethanesulphonyl)imide, Bis(trifluoromethanesulphonyl)imide manganese(II) salt

M2862-1G M2862-5G
TCIAM2862-1GEA 74 EUR
TCIAM2862-1G TCIAM2862-5G
Manganesebis(trifluoromethylsulphonyl)imide ≥97.0% (por análise titulométrica)
Manganesebis(trifluoromethylsulphonyl)imide
Fórmula: C₄F₁₂MnN₂O₈S₄
Peso Molecular: 615.21 g/mol
Número MDL : MFCD23380173
Número CAS: 207861-55-0
UN: 1759
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c06aa6e065688955ca592888ba6349e8b1f4c2bc&vid=c06aa6e065688955ca592888ba6349e8b1f4c2bc&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 97.0 %
Purity (Ion exchange titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR