Print…

3,3'-Methylenebis(1-tert-butyl-3-imidazolium bromide) ≥98.0% (por base de azoto total)

Fornecedor: TCI
Atenção

M2790-1G M2790-5G
TCIAM2790-1GEA 66 EUR
TCIAM2790-1G TCIAM2790-5G
3,3'-Methylenebis(1-tert-butyl-3-imidazolium bromide) ≥98.0% (por base de azoto total)
3,3'-Methylenebis(1-tert-butyl-3-imidazolium bromide)
Fórmula: C₁₅H₂₆Br₂N₄
Peso Molecular: 422.21 g/mol
Número CAS: 247902-48-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96e260c956ecfe0ff35c123aaec1441ab0d04ca3&vid=96e260c956ecfe0ff35c123aaec1441ab0d04ca3&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR