Print…

Methyl-β-D-ribofuranoside ≥97.0% (por GC)

Fornecedor: TCI

Sinônimos: (2R,3S,4R,5R)-2-(Hydroxymethyl)-5-methoxytetrahydrofuran-3,4-diol, Methyl β-D-Ribofuranoside

M1965-1G M1965-5G
TCIAM1965-1GEA 31 EUR
TCIAM1965-1G TCIAM1965-5G
Methyl-β-D-ribofuranoside ≥97.0% (por GC)
Methyl-beta-D-ribofuranoside
Fórmula: C₆H₁₂O₅
Peso Molecular: 164.16 g/mol
Número MDL : MFCD00047075
Número CAS: 7473-45-2
EINECS: 231-271-5

Encomendar

Especificações dos Resultados do Teste

Appearance White to Almost white powder to crystaline
Purity (GC) min. 97.0 %
Melting point 76.0 to 80.0 deg-C
Specific rotation [a]20/D -45.0 to -55.0 deg (C=2, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR