Print…

Methyl-2-(p-tolyl)benzoate ≥98.0% (por GC)

Fornecedor: TCI

Sinônimos: Methyl 2-(4-Methylphenyl)benzoate, 4'-Methylbiphenyl-2-carboxylic Acid Methyl Ester, Methyl 4'-methyl-[1,1'-biphenyl]-2-carboxylate, Methyl 4'-Methylbiphenyl-2-carboxylate, 2-(p-Tolyl)benzoic Acid Methyl Ester

M1809-25G M1809-5G
TCIAM1809-25GEA 132 EUR
TCIAM1809-25G TCIAM1809-5G
Methyl-2-(p-tolyl)benzoate ≥98.0% (por GC)
Methyl-2-(p-tolyl)benzoate
Fórmula: C₁₅H₁₄O₂
Número MDL : MFCD03453661
Número CAS: 114772-34-8

Encomendar

Especificações dos Resultados do Teste

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR