2-Methylbenzo[e]indene (mixture of 1H- and 3H-form) ≥97.0% (por GC)
Fornecedor: TCI
M1425-1G
M1425-100MG
TCIAM1425-1GEA
847
EUR
TCIAM1425-1G
TCIAM1425-100MG
2-Methylbenzo[e]indene (mixture of 1H- and 3H-form) ≥97.0% (por GC)
2-Methylbenzo[e]indene (mixture of 1H- and 3H-form)
|
Especificações dos Resultados do Teste
Appearance | Yellow to Amber clear liquid |
Purity (GC) | min. 97.0 % (total of isomers) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...