Print…

Methyl nonafluorobutyl ether ≥98.0% (por GC)

Fornecedor: TCI

Sinônimos: Perfluorobutyl Methyl Ether, Nonafluorobutyl Methyl Ether, Methyl Perfluorobutyl Ether

M1345-25G M1345-500G
TCIAM1345-25GEA 18 EUR
TCIAM1345-25G TCIAM1345-500G
Methyl nonafluorobutyl ether ≥98.0% (por GC)
Methyl nonafluorobutyl ether
Fórmula: C₅H₃F₉O
Peso Molecular: 250.06 g/mol
Número MDL : MFCD00674093
Número CAS: 163702-07-6

Encomendar

Especificações dos Resultados do Teste

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR