Print…

Methyl-β-D-xylopyranoside ≥96.0% (por GC)

Fornecedor: TCI

M1253-25G M1253-5G
TCIAM1253-25GEA 185 EUR
TCIAM1253-25G TCIAM1253-5G
Methyl-β-D-xylopyranoside ≥96.0% (por GC)
Methyl-beta-D-xylopyranoside
Fórmula: C₆H₁₂O₅
Número MDL : MFCD00047532
Número CAS: 612-05-5
EINECS: 210-289-7

Encomendar

Especificações dos Resultados do Teste

Appearance White to Light yellow powder to crystal
Purity (GC) min. 96.0 %
Specific rotation [a]20/D -60.0 to -65.0 deg (C=2, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR