Print…

Lanthanum(III) bis(trifluoromethanesulphonyl)imide ≥98.0% (por análise titulométrica)

Fornecedor: TCI
Perigo

Sinônimos: Bis(trifluoromethanesulphonyl)imide Lanthanum(III) salt, Lanthanum(III) triflimide

L0294-1G L0294-5G
TCIAL0294-1GEA 62 EUR
TCIAL0294-1G TCIAL0294-5G
Lanthanum(III) bis(trifluoromethanesulphonyl)imide ≥98.0% (por análise titulométrica)
Lanthanum(III) bis(trifluoromethanesulphonyl)imide
Fórmula: C₆F₁₈LaN₃O₁₂S₆
Número CAS: 168106-26-1
UN: 1759
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3bd080d7deaf2193e8e0dac47fa2cf178799f156&vid=3bd080d7deaf2193e8e0dac47fa2cf178799f156&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 %
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR