Print…

Galvinoxyl radical livre

Fornecedor: TCI

G0020-1G G0020-5G
TCIAG0020-1GEA 126 EUR
TCIAG0020-1G TCIAG0020-5G
Galvinoxyl radical livre
Galvinoxyl
Fórmula: C₂₉H₄₁O₂
Temperatura de Armazenamento: Frigorífico
Número MDL : MFCD00001616
Número CAS: 2370-18-5
EINECS: 219-133-2
Merck Index: 14,04357

Encomendar

Especificações dos Resultados do Teste

Appearance Dark red to Dark purple to Dark blue powder to crystal
Lambda max. 287.0 to 301.0 nm (Isooctane)
Lambda max.2 398.0 to 402.0 nm (Isooctane)
Lambda max.3 421.0 to 425.0 nm (Isooctane)
Molar extinction coefficient min. 9,400 (Isooctane, 287.0 to 301.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR