Print…

Hexadecyl gallate ≥95.0% (por GC)

Fornecedor: TCI

Sinônimos: Gallic Acid Cetyl Ester, Cetyl Gallate, Gallic Acid Hexadecyl Ester

G0013-25G G0013-5G
TCIAG0013-25GEA 285 EUR
TCIAG0013-25G TCIAG0013-5G
Hexadecyl gallate ≥95.0% (por GC)
Hexadecyl gallate
Fórmula: C₂₃H₃₈O₅
Número MDL : MFCD00016428
Número CAS: 5026-65-3
EINECS: 225-715-7

Encomendar

Especificações dos Resultados do Teste

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Melting point 98.0 to 101.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR