Print…
Farnesyl acetate (mixture of isomers) ≥95.0% (por GC)
Fornecedor: TCI
Sinônimos: 3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol Acetate
F0347-25ML F0347-5ML
TCIAF0347-25MLEA 59 EUR
TCIAF0347-25ML TCIAF0347-5ML
Farnesyl acetate (mixture of isomers) ≥95.0% (por GC)
Farnesyl acetate (mixture of isomers)
Fórmula: C₁₇H₂₈O₂
Peso Molecular: 264.41 g/mol
Ponto de Inflamação: 95°C
Número MDL : MFCD00036516
Número CAS: 29548-30-9
EINECS: 249-689-1
Encomendar
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 % (total of isomer)