Dodecaethylene glycol monomethyl ether ≥90.0% (por GC)
Fornecedor: TCI
Sinônimos:
mPEG12-Alcohol, m-PEG12-OH
D2904-100MG
D2904-1G
TCIAD2904-100MGEA
90
EUR
TCIAD2904-100MG
TCIAD2904-1G
Dodecaethylene glycol monomethyl ether ≥90.0% (por GC)
Dodecaethylene glycol monomethyl ether
Fórmula:
C₂₅H₅₂O₁₃ Peso Molecular: 560.68 g/mol Temperatura de Armazenamento: Congelador |
Número MDL :
MFCD06797088 Número CAS: 5702-16-9 |
Especificações dos Resultados do Teste
Appearance | White or Colorles to Yellow to Orange powder to lump to clear liquid |
Purity (GC) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...