Print…

1,3-Dicyclohexylbarbituric acid ≥98.0% (por base de azoto total)

Fornecedor: TCI

D2815-1G D2815-5G
TCIAD2815-1GEA 74 EUR
TCIAD2815-1G TCIAD2815-5G
1,3-Dicyclohexylbarbituric acid ≥98.0% (por base de azoto total)
1,3-Dicyclohexylbarbituric acid
Fórmula: C₁₆H₂₄N₂O₃
Peso Molecular: 292.38 g/mol
Número MDL : MFCD01216967
Número CAS: 35824-91-0

Encomendar

Especificações dos Resultados do Teste

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 202.0 to 206.0 deg-C
Solubility in hot Acetone almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR