Dehydroepiandrosterone ≥99.0% (por GC)
Fornecedor: TCI
D0044-1G
D0044-25G
D0044-5G
TCIAD0044-1GEA
25
EUR
TCIAD0044-1G
TCIAD0044-25G
TCIAD0044-5G
Dehydroepiandrosterone ≥99.0% (por GC)
Dehydroepiandrosterone
Fórmula:
C₁₉H₂₈O₂ |
Número MDL :
MFCD00003613 Número CAS: 53-43-0 EINECS: 200-175-5 Merck Index: 14,02871 |
Especificações dos Resultados do Teste
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 148.0 to 152.0 deg-C |
Specific rotation [a]20/D | +11.5 to +13.5 deg (C=2, EtOH) |
Solubility in EtOH | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...