Print…

Cholesterol butyrate ≥95.0% (por GC)

Fornecedor: TCI

Sinônimos: Butyric acid cholesterol ester

C0668-25G C0668-500G
TCIAC0668-25GEA 68 EUR
TCIAC0668-25G TCIAC0668-500G
Cholesterol butyrate ≥95.0% (por GC)
Cholesterol butyrate
Fórmula: C₃₁H₅₂O₂
Peso Molecular: 456.76 g/mol
Número MDL : MFCD00021144
Número CAS: 521-13-1
EINECS: 208-304-7

Encomendar

Especificações dos Resultados do Teste

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Specific rotation [a]20/D -28.0 to -32.0 deg (C=2, THF)
Solubility in THF very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR