tert-Butyl triphenylphosphoranylidenecarbamate ≥98.0% (por HPLC, azoto total)
Fornecedor: TCI
Sinônimos:
N-Boc-imino(triphenyl)phosphorane, (Triphenylphosphoranylidene)carbamic acid tert-Butyl ester, N-Boc-Imino-(triphenyl)phosphorane, tert-Butyl (Triphenylphosphoranylidene)carbamate, N-(tert-Butoxycarbonyl)imino(triphenyl)phosphorane
B5687-5G
B5687-25G
TCIAB5687-5GEA
50
EUR
TCIAB5687-5G
TCIAB5687-25G
tert-Butyl triphenylphosphoranylidenecarbamate ≥98.0% (por HPLC, azoto total)
tert-Butyl triphenylphosphoranylidenecarbamate
Fórmula:
C₂₃H₂₄NO₂P |
Número MDL :
MFCD00798174 Número CAS: 68014-21-1 |
|
Especificações dos Resultados do Teste
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 147.0 to 151.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...