Print…

N,N'-Bis[4-(diphenylamino)phenyl]-N,N'-diphenylbenzidine ≥98.0% (por HPLC)

Fornecedor: TCI

B5117-200MG B5117-1G
TCIAB5117-200MGEA 63 EUR
TCIAB5117-200MG TCIAB5117-1G
N,N'-Bis[4-(diphenylamino)phenyl]-N,N'-diphenylbenzidine ≥98.0% (por HPLC)
N,N'-Bis[4-(diphenylamino)phenyl]-N,N'-diphenylbenzidine
Fórmula: C₆₀H₄₆N₄
Número CAS: 209980-53-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63adbc2392f3a59e3f61e1f356d204bb5b02b99f&vid=63adbc2392f3a59e3f61e1f356d204bb5b02b99f&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 248.0 to 252.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR