Print…

Betaxolol hydrochloride ≥98.0% (por HPLC)

Fornecedor: TCI
Atenção

B4474-50MG B4474-250MG
TCIAB4474-50MGEA 97 EUR
TCIAB4474-50MG TCIAB4474-250MG
Betaxolol hydrochloride ≥98.0% (por HPLC)
Betaxolol hydrochloride
Fórmula: C₁₈H₂₉NO₃
Peso Molecular: 343.89 g/mol
Número MDL : MFCD00242959
Número CAS: 63659-19-8
EINECS: 264-384-3
Merck Index: 14,01182

Encomendar

Especificações dos Resultados do Teste

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 113.0 to 117.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR