Print…

2-Benzylbenzimidazole hydrochloride ≥98.0% (por HPLC)

Fornecedor: TCI
Atenção

B4341-5G B4341-25G
TCIAB4341-5GEA 88 EUR
TCIAB4341-5G TCIAB4341-25G
2-Benzylbenzimidazole hydrochloride ≥98.0% (por HPLC)
2-Benzylbenzimidazole hydrochloride
Fórmula: C₁₄H₁₂N₂
Número MDL : MFCD00443659
Número CAS: 1212-48-2
EINECS: 214-921-2

Encomendar

Especificações dos Resultados do Teste

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Purity (Argentmetric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR