Isobutyl butyrate, Sigma-Aldrich®
Fornecedor: Merck
Sinônimos:
Butyric acid Isobutyl ester, 2-Methyl-1-propylbutyrate
Nível de qualidade M-Clarity™ = MQ300
W218707-1KG-K
W218707-9KG-K
SIALW218707-1KG-KEA
83.2
EUR
SIALW218707-1KG-K
SIALW218707-9KG-K
Isobutyl butyrate, Sigma-Aldrich®
Isobutyl butyrate
Fórmula:
C₈H₁₆O₂ Peso Molecular: 144.21 g/mol |
Número MDL :
MFCD00027134 Número CAS: 539-90-2 EINECS: 208-729-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...