Print…

Aegeline ≥98.0% (por HPLC, azoto total)

Fornecedor: TCI

Sinônimos: Aegelin, N-(2-Hydroxy-2-(4-methoxyphenyl)ethyl)cinnamamide, (E)-N-[2-Hydroxy-2-(4-methoxyphenyl)ethyl]cinnamamide

A3170-5G A3170-25G
TCIAA3170-5GEA 43 EUR
TCIAA3170-5G TCIAA3170-25G
Aegeline ≥98.0% (por HPLC, azoto total)
Aegeline
Fórmula: C₁₈H₁₉NO₃
Número MDL : MFCD00048045
Número CAS: 456-12-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec51db07bd1aea1502001659cea8525d2d38773c&vid=ec51db07bd1aea1502001659cea8525d2d38773c&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 177.0 to 181.0 deg-C
Specific rotation [a]20/D +49.0 to +53.0 deg (C=0.01, CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR