Diethyl acetonylmalonate ≥95.0% (por GC)
Fornecedor: TCI
A1503-1G
A1503-5G
TCIAA1503-1GEA
50
EUR
TCIAA1503-1G
TCIAA1503-5G
Diethyl acetonylmalonate ≥95.0% (por GC)
Diethyl acetonylmalonate
Fórmula:
C₁₀H₁₆O₅ |
Número MDL :
MFCD00177949 Número CAS: 23193-18-2 |
Especificações dos Resultados do Teste
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...