Praseodymium trichloride hydrate, Sigma-Aldrich®
Fornecedor: Merck
Sinônimos:
Praseodymium (III) chloride hydrate
Nível de qualidade M-Clarity™ = MQ100
205141-10G
205141-50G
SIAL205141-10GEA
118
EUR
SIAL205141-10G
SIAL205141-50G
Praseodymium trichloride hydrate, Sigma-Aldrich®
Praseodymium trichloride hydrate
Fórmula:
Cl₃Pr·H₂O |
Número MDL :
MFCD00149828 Número CAS: 19423-77-9 EINECS: 233-794-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...