Print…
tert-Butylchlorodimethylsilane 50% em tolueno, Sigma-Aldrich®
Fornecedor: Merck
Sinônimos: tert-Butyldimethylsilyl Chloride, Chloro-tert-butyldimethylsilane, t-Butyldimethylchlorosilane, tert-Butyldimethylchlorosilane, tert-Butyldimethylsilyl chloride, TBDMS chloride
Nível de qualidade M-Clarity™ = MQ200
473464-100G 473464-50G
SIAL473464-100GEA 241 EUR
SIAL473464-100G SIAL473464-50G
tert-Butylchlorodimethylsilane 50% em tolueno, Sigma-Aldrich®
tert-Butylchlorodimethylsilane
Fórmula: C₆H₁₅ClSi
Número MDL : MFCD00000501
Número CAS: 18162-48-6
EINECS: 242-042-4
UN: 2924
ADR: 3,II