Print…
Human Recombinant IL-1B (Animal free) (from E. coli)
Fornecedor: Shenandoah Biotechnology
100-167AF-100U 100-167AF-10UG 100-167AF-1MG 100-167AF-500U
SHBT100-167AF-100UEA 572 EUR
SHBT100-167AF-100U SHBT100-167AF-10UG SHBT100-167AF-1MG SHBT100-167AF-500U
Human Recombinant IL-1B (Animal free) (from E. coli)
Proteínas e Peptídeos
Interleukin 1 beta (IL-1β) is a pro-inflammatory cytokine that is produced by monocytes, macrophages, and dendritic cells (DCs).  IL-1β signals through the IL-1 receptor, type 1 (IL-1R1) to activate the myeloid differentiation primary response 88 (MYD88) signaling pathway, which contains the cytoplasmic Toll/IL-1 receptor (TIR) domain adapter.
  • High quality
  • Low endotoxin
  • Affordable
IL-1β and the independently regulated IL-1α protein have overlapping pro-inflammatory activities to induce adhesion molecule expression on epithelial cells, control fever induction, initiate rheumatoid arthritis, and promote septic shock.
Fornecimento: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Atenção: For research use only.