Print…
Safety shoes, Low ESD S3, GHOST BOA®
Fornecedor: ELTEN
72081/40 72081/41 72081/46 72081/44 72081/35 72081/36 72081/37 72081/38 72081/39 72081/47 72081/48 72081/43 72081/45 72081/42
114-8676EA 94.9 EUR
114-8676 114-8677 114-8682 114-8680 114-8671 114-8672 114-8673 114-8674 114-8675 114-8683 114-8684 114-8679 114-8681 114-8678
Safety shoes, Low ESD S3, GHOST BOA®
Sapatos e botas
Designed for indoor or outdoor use, GHOST BOA® safety shoes are ideally suited for areas exposed to moisture, areas with a risk of penetration from pointed and sharp objects, or areas with a risk of electrostatic discharge.
  • Hydrophobized microfibre upper
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay ESD PRO sole
  • Non-metallic, penetration-resistant midsole
  • Abrasion-proof toe protection
  • Steel toe cap
  • Excellent slip resistance
Offering excellent wearing comfort, the padded upper edge protects the Achilles tendon. A full padded bellows tongue prevents pressure marks and deters dirt from entering into the shoe. The BOA® Fit closure system allows them to be opened/closed with just one hand, without the need for laces, buckles, or velcro. The full-length, exchangeable inlay ESD PRO sole provides the highest possible comfort in safety shoes. The inlay sole is functionally absorbing and releasing moisture, so the foot is always kept comfortably dry.
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). The shoes comply to the standard 61340-5-1.
Certificações: EN ISO 20345 S3.