Print…
N,O-Bis(trimethylsilyl)trifluoroacetamide with 10% trimethylchlorosilane 98% (excluding TMCS) (por GC), LiChropur™ para derivatização GC, Supelco®
Fornecedor: Merck
Perigo
Nível de qualidade M-Clarity™ = MQ100
15209-5ML 15209-25ML 15209-10X1ML
15209-5MLEA 50.8 EUR
15209-5ML 15209-25ML 15209-10X1ML
N,O-Bis(trimethylsilyl)trifluoroacetamide with 10% trimethylchlorosilane 98% (excluding TMCS) (por GC), LiChropur™ para derivatização GC, Supelco®
N,O-Bis(trimethylsilyl)trifluoroacetamide with 10% trimethylchlorosilane
Fórmula: C₈H₁₈F₃NOSi₂
Peso Molecular: 257.4 g/mol
Número MDL : MFCD00008269
Número CAS: 25561-30-2
UN: 2924
ADR: 8(3) II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83130879a8837570320458cc2c72db1f8eee25a3&vid=27b1e6737d4568323c7ec921ad3c7fde50b835b6&prid=web&clid=SAPDAM