Print…
HMDS (1,1,1,3,3,3-hexametildisilazano) ≥99.0% (by GC), LiChropur™ for GC derivatisation, Supelco®
Fornecedor: Merck
52619-250ML 52619-50ML
52619-250MLEA 288 EUR
52619-250ML 52619-50ML
HMDS (1,1,1,3,3,3-hexametildisilazano) ≥99.0% (by GC), LiChropur™ for GC derivatisation, Supelco®
HMDS (1,1,1,3,3,3-hexamethyldisilazane)
Fórmula: (CH₃)₃SiNHSi(CH₃)₃
Ponto de Fusão: –82 °C
Número MDL : MFCD00008259
Número CAS: 999-97-3
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bb897a49260bc7677ad1f6d36b4d92877583818&vid=aacae6d820fe7f24349e379841e4cf581b4c637f&prid=web&clid=SAPDAM