Print…
Precision balances, Cubis II series
Fornecedor: Sartorius Balances
MCA1203SE-CEU MCA1203SE-S00 MCA1203SR-CEU MCA1203SR-S00 MCA2202S0-CEU MCA2202S0-S00 MCA2203SE-CEU MCA2203SE-S00 MCA2203SR-CEU MCA2203SR-S00 MCA3203SE-CEU MCA3203SE-S00 MCA3203SR-CEU MCA3203SR-S00 MCA4202S0-CEU MCA4202S0-S00 MCA5203SE-CEU MCA5203SE-S00 MCA5203SR-CEU MCA5203SR-S00 MCA6202S0-CEU MCA623SE-CEU MCA623SE-S00 MCA623SR-CEU MCA623SR-S00 MCA8201S0-CEU MCA8201S0-S00 MCE1203SR-CEU MCE1203SR-S00 MCE2202S0-CEU MCE2202S0-S00 MCE2203SR-CEU MCE2203SR-S00 MCE3203SR-CEU MCE3203SR-S00 MCE4202S0-CEU MCE4202S0-S00 MCE6202S0-CEU MCE6202S0-S00 MCE623SR-CEU MCE623SR-S00 MCE8201S0-CEU MCE8201S0-S00 MCA6202S0-S00
611-4681EA 12732 EUR
611-4681 611-4682 611-4683 611-4684 611-4699 611-4700 611-4701 611-4702 611-4703 611-4704 611-4715 611-4716 611-4717 611-4718 611-4727 611-4728 611-4729 611-4730 611-4731 611-4732 611-4741 611-4743 611-4744 611-4745 611-4746 611-4749 611-4750 611-4751 611-4752 611-4761 611-4762 611-4763 611-4764 611-4771 611-4772 611-4781 611-4782 611-4783 611-4784 611-4785 611-4786 611-4787 611-4788 611-4742
Precision balances, Cubis II series
Balanças
The Cubis® II range has a modular design that to allows the customer to choose the components that suit the application area that is of interest. For the customer, this configurable concept delivers quality weighing, together with a high degree of flexibility, without having to invest in expensive equipment that has features that may never be required.
  • Quick and easy motorised automatic levelling, for weighing modules up to 8,2 kg, units above 8,2 kg have manual levelling
  • Gesture control sensor recognises 4 gestures, it allows user to send a document to a device
  • Connectivity package links to diverse laboratory information systems (Windows File Server (SMB), FTP, FTPS and STARLIMS) without the need for additional software
  • Full data integrity following ALCOA+ principles, the balances audit trail is a tamper protected, time stamped electronic logfile traceable to UTC and the balance contains an Alibi memory
Two different user interfaces are available to cover all facets of laboratory applications and enable a range of options for balance operation. The customer can then add the appropriate weighing platform to the choice of control unit.
MCE Essential User Interface: This is a large, high contrast touch display with factory installed essential weighing applications for easy operation.
MCA Advanced User Interface: This is a large 7" colour touch display featuring touch, scroll and swipe functionality with factory installed essential weighing applications and diverse QApp packages for optional software extension. An optional climate sensor which monitors the influencing environmental variables ensuring temperature, pressure and humidity do not negatively affect results is available. The Status Center shows all information about the balance and environmental conditions (calibration, levelling, temperature, humidity, air pressure and service) in a dashboard, giving detailed help and support in the event of warnings or errors.
Various Q-Apps, customer-specific applications, can be integrated into the balance. The applications are used to create customer-specific weighing tasks. These are reloadable application programs that guide the user step-by-step through the specific workflow, guaranteeing that the procedures described in the corresponding SOPs are observed at all times. As individual Q-Apps are performed according to their specific application, a variety of solutions for differential weighing, formula and filling quantity control applications are available as Q-App packages. Workflows can be performed independently and do not necessarily require a PC connection.
Certificações: Supports all requirements for FDA directive 21 CFR part II and EU Annex II compliance.
Fornecimento: There are different optional software packages, each contains applications or functions to enhance and customise the balance for the users’ workflows. Choice of QApp Package: Pharma; Advanced Applications: Utilities; Connectivity or the all inclusive QApps package includes all of these 4 packages. (denoted by QP99).
Atenção: Note: Models with suffix -CEU are verified version Europe (except FR) and -S00 suffix denotes standard version worldwide.