Print…
Cut resistant gloves, uvex NK 4022
Fornecedor: UVEX
6020203 6020204
UVEX6020203EA 470 EUR
UVEX6020203 UVEX6020204
Cut resistant gloves, uvex NK 4022
Luvas
These cotton/aramid safety gloves with NBR coating provide reliable and durable cut protection. The rough surface provides a very good grip, while the glove also offers heat protection.
  • Suitable for contact heat up to +100 °C
  • Very good tear and abrasion resistance
  • Very good grip in dry, damp and oily areas
  • Good thermal insulation in direct contact with warm to hot objects
  • Comfortable fit
Areas of application: Heavy-duty mechanical work, sheet metal processing, machinery and tool manufacturing, handling cold or hot objects.
Certificações: EN 388:2016 (2342X), EN 407 (X1XXXX)