RNasin® Plus ribonuclease inhibitor
Fornecedor: Promega
RNasin® Ribonuclease inhibitors are widely used protection against RNases worldwide.
- Use in many downstream assays: Inhibits a broad spectrum of eukaryotic RNases over a wide pH range (pH 5 to 8)
- Maintains inhibitory activity over a wide temperature range
- Does not inhibit SP6, T7 or T3 RNA Polymerase; GoScript™ Reverse Transcriptase, AMV or M-MLV Reverse Transcriptase; or Taq DNA polymerase
RNasin protects RNA from degradation by RNases A, B and C, bringing you peace of mind when working with RNA from valuable source materials. Use RNasin® Ribonuclease Inhibitor for RNA isolation, RT-PCR, in vitro transcription or any situation where RNase is a concern.
Available as native and recombinant forms of RNAsin® Inhibitor, as well as RNAsin® Plus an enhanced formulation provides increased resistance to oxidation and stability up to 70 °C.