Print…

Amylose (de batata), Sigma-Aldrich®

Fornecedor: Merck
Atenção


Nível de qualidade M-Clarity™ = MQ300

A0512-1G A0512-250MG A0512-25G A0512-5G
SIALA0512-1GEA 164 EUR
SIALA0512-1G SIALA0512-250MG SIALA0512-25G SIALA0512-5G
Amylose (de batata), Sigma-Aldrich®
Amylose

Amylose is a polyglucose polysaccharide which together with amylopectin makes starch(s).


The glucosidic bonds of amylopectin are α-1,4 linkages. Amylose is used to identify, differentiate and characterise amylase(s).


Amylose is used as a thickening agent, a gelling agent and emulsion stabiliser. Amylose is used in the preparation of films and membranes with potential use in drug delivery.

Fórmula: (C₆H₁₀O₅)n
Temperatura de Armazenamento: Ambiente
Número MDL : MFCD00134652
Número CAS: 9005-82-7

Encomendar

Especificações dos Resultados do Teste

Solvent Content (by GC) as Ethanol ≤10%
Solvent Content (by GC) as Butanol ≤2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR