Print…

Ytterbium(III) iodide, Sigma-Aldrich®

Fornecedor: Merck

Sinônimos: Ytterbium triiodide, Ytterbium(II) iodide

Nível de qualidade M-Clarity™ = MQ100

494372-1G 494372-5G
SIAL494372-1GEA 443 EUR
SIAL494372-1G SIAL494372-5G
Ytterbium(III) iodide, Sigma-Aldrich®
Ytterbium(III) iodide
Fórmula: YbI₃
Número CAS: 13813-44-0
EINECS: 237-474-5

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR