Print…

Octadecyltrimethoxysilane, Sigma-Aldrich®

Fornecedor: Merck

Sinônimos: Trimethoxy(octadecyl)silane

Nível de qualidade M-Clarity™ = MQ100

376213-25ML 376213-5ML
SIAL376213-25MLEA 234 EUR
SIAL376213-25ML SIAL376213-5ML
Octadecyltrimethoxysilane, Sigma-Aldrich®
Octadecyltrimethoxysilane
Fórmula: C₂₁H₄₆O₃Si
Peso Molecular: 374.68 g/mol
Número MDL : MFCD00043060
Número CAS: 3069-42-9
EINECS: 221-339-2

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR